Learn More
ARHGAP40 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560145
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AGKWKAAETR LFGVPLDSLL EADHKVLPST QVPLVLQALL SCLEKRGLDM EGILRVPGSQ ARVKGLEQKL ERDFYAGLFS WDEVHHNDAS DLLKRFIR Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.Specifications
ARHGAP40 | |
Polyclonal | |
Unconjugated | |
ARHGAP40 | |
ARHGAP40; C20orf95; dJ1100H13.4; Rho GTPase activating protein 40; rho GTPase-activating protein 40; rho-type GTPase-activating protein 40 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
343578 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TG30 | |
ARHGAP40 | |
Recombinant protein corresponding to Human ARHGAP40 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only