Learn More
ARHGEF11 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30508-1-AP-20UL
Description
The ARHGEF11 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ARHGEF11. This antibody recognizes human, mouse antigen. The ARHGEF11 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| ARHGEF11 | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| ARHGEF11, DKFZp667F1223, GTRAP48, KIAA0380, PDZ RHOGEF | |
| 20 μL | |
| 9826 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |