missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL4C Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10202-1-AP-150UL
This item is not returnable.
View return policy
Description
ADP-ribosylation factor-like 4C is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4C is closely similar to ARL4A and ARL4D and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in cholesterol transport.Specifications
| ARL4C | |
| Polyclonal | |
| Unconjugated | |
| ARL4C | |
| ARL4C, ARL7, LAK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10123 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P56559 | |
| ARL4C | |
| ARL4C Fusion Protein Ag0246 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction