missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMC12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21386-100ul
This item is not returnable.
View return policy
Description
ARMC12 Polyclonal antibody specifically detects ARMC12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ARMC12 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| armadillo repeat containing 12, C6orf81, chromosome 6 open reading frame 81, FLJ25390, hypothetical protein LOC221481 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: QYLGQLDIRKSVVSLATGAGAIYLLYKAIKAGIKCKPPLCSNSPICIARLAVERERHGRDSGELRRLLNSLECKQDEYA | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 221481 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction