missing translation for 'onlineSavingsMsg'
Learn More

ARPC1A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

216.00 € - 539.00 €

Specifications

Antigen ARPC1A
Dilution Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, KnockDown Validated
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18421221
View Documents
Novus Biologicals
NBP1-88851-25ul
25 μL
216.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18251347
View Documents
Novus Biologicals
NBP1-88851
0.1 mL
539.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ARPC1A Polyclonal specifically detects ARPC1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

ARPC1A
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
actin binding protein (Schizosaccharomyces pombe sop2-like), actin related protein 2/3 complex, subunit 1A (41 kD), actin related protein 2/3 complex, subunit 1A, 41kDa, actin-related protein 2/3 complex subunit 1A, SOP2Hs, SOP2LArc40, SOP2-like protein
ARPC1A
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, KnockDown Validated
Polyclonal
Rabbit
Human
Q92747
10552
This antibody was developed against Recombinant Protein corresponding to amino acids:HGVSFSASGSRLAWVSHDSTVSVADASKSVQVSTLKTEFLPLLSVSFVSENSVVAAGHDCCPMLFNYDDRGCLTFVSKLDIPKQSIQRNMSAMERFRNMDKRATTEDRNTALETLHQNSITQVSIYEVDKQDCRKFCTTGI
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
42 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.