Learn More
ARRDC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21030-1-AP-20UL
This item is not returnable.
View return policy
Description
The ARRDC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ARRDC1. This antibody recognizes human antigen. The ARRDC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ARRDC1 | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| ARRDC1, arrestin domain containing 1 | |
| 20 μL | |
| 92714 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |