missing translation for 'onlineSavingsMsg'
Learn More

Arsenazo III

Product Code. p-7042581
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15465299

Brand: Thermo Scientific Alfa Aesar J63031.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Arsenazo III (3,6-bis((E)-(2-Arsonophenyl)diazenyl)-4,5-dihydroxynaphthalene-2,7-disulfonic acid, CAS # 1668-00-4) is a metallochrome indicator that is used for detecting calcium levels in plasma and serum. | CAS: 1668-00-4 | C22H18As2N4O14S2 | 776.363 g/mol

Suitable for determination of micromolar amounts of calcium

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Arsenazo III is highly specific for calcium and produces absorbance peaks in the 600-660 nm range
• It reacts in slightly acidic conditions, and is soluble in alcohol and semi-soluble in water

Application

• Arsenazo III can detect calcium in permeabilized cells; it is also capable of detecting uranium and thorium
• Arsenazo III has been used in vitro to measure intracellular Ca2+ levels in live neurons alongside patch clamp methods

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1668-00-4
Molecular Formula C22H18As2N4O14S2
Molecular Weight (g/mol) 776.363
MDL Number MFCD00036695
InChI Key TVMZRHVOFZTNET-RIRMOVKSSA-N
Synonym arsenazo iii, arsenazo iii sodium salt, 2,2'-1, 8-dihydroxy-3,6-disulfo-2, 7-naphthalene-bis azo dibenzenearsonic acid, 2,7-bis 2-arsonophenylazo-1, 8-dihydroxy-3, 6-naphthalene disulfonic acid, 2,7-bis 2-arsonophenylazo-1,8-dihydroxynaphthalene-3,6-disulfonic acid, 2,7-bis 2-arsonophenylazo chromotropic acid, 2,2'-1,8-dihydroxy-3,6-disulfonaphthylene-2,7-bisazo bisbenzenearsonic acid, 3,6-bis 2-2-arsonophenyl diazen-1-yl-4,5-dihydroxynaphthalene-2,7-disulfonic acid, 3,6-bis 2-2-arsonophenyl diazenyl-4,5-dihydroxy-2,7-naphthalenedisulfonic acid, 3,6-bis 2-2-arsonophenyl hydrazin-1-ylidene-4,5-dioxonaphthalene-2,7-disulfonic acid
PubChem CID 9810878
IUPAC Name (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O

Specifications

Color Brown to Purple
Quantity 5 g
UN Number UN3465
Beilstein 5717957
Formula Weight 776.37
Physical Form Powder
Chemical Name or Material Arsenazo III

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.