missing translation for 'onlineSavingsMsg'
Learn More

Arsenic for AAS 0.02mg/L matrix HNO3 diluted, Fisher Chemical™
BIOPHARMA

Product Code. 12984525 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12984525 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12984525 Supplier Fisher Chemical Supplier No. E 003 050
  • ONLINE EXCLUSIVE PRICE
    106.00 €
    81.34 € / 50mL
    Save 24.66 €
    23% Off
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 7697-37-2
Physical Form Liquid
Molecular Formula HNO3
Concentration 0.02 mg/L
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Type Flame Single-element Standard Solution
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Solvent or Matrix 1% HNO3 (diluted)
Molecular Weight (g/mol) 63.01
PubChem CID 944
Quantity 50 mL
ChEBI CHEBI:48107
Chemical Name or Material Arsenic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.