missing translation for 'onlineSavingsMsg'
Learn More

Arsenic III Speciation Standard (Assurance™ Grade/For ICP-MS), SPEX CertiPrep™

Product Code. 10394433 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Packaging:
Poly Bottle
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10394433 125 mL Poly Bottle 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10394433 Supplier SPEX Certiprep Supplier No. SPECAS3
  • ONLINE EXCLUSIVE PRICE
    78.81 €
    70.02 € / 125mL
    Save 8.79 €
    11% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-38-2 | As | 74.92 g/mol

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Arsenic +3
  • Speciation Standard
  • 125 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Chemical Identifiers

CAS 7440-38-2 , 7647-01-0 , 7732-18-5
Molecular Formula As
Molecular Weight (g/mol) 74.92
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
PubChem CID 5359596
ChEBI CHEBI:27563
IUPAC Name arsenic
SMILES [As]

Specifications

Packaging Poly Bottle
CAS Min % 0.1, 2.0, 97.9
CAS Max % 0.1, 2.0, 97.9
Technique ICP-MS
Solution Type Standard Solution
Type ICP/ICP-MS Standard
Quantity 125 mL
Grade Assurance
Chemical Name or Material Arsenic III Speciation Standard
compliance-icons
Product Identifier
  • 1000 ug/mL Arsenic (III)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid, Arsenic from Arsenictrioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.