missing translation for 'onlineSavingsMsg'
Learn More

Arsenic Standard: 1000 μg/g As in 75 cSt Hydrocarbon Oil, VHG™

Product Code. 17981636
Change view
Click to view available options
Quantity:
50 g
200 g
400 g
Unit Size:
200g
400g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17981636 400 g 400g
17900936 200 g 200g
17941606 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17981636 Supplier VHG Labs Supplier No. VHGOAS1000400G
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Arsenic Standard: 1000 μg/g As in 75 cSt Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Arsenic
CAS 7440-38-2
Methods ICP/XRF/RDE
Analyte or Component Names Arsenic
Shipping Condition Room Temperature
Molecular Formula As
Certifications/Compliance ISO 17034
Synonym Arsenicals, Arsenic, inorganic, Arsen
InChI Formula InChI=1S/As
SMILES [As]
IUPAC Name arsenic
Solvent or Matrix 75 cSt Mineral Oil
Molecular Weight (g/mol) 74.92
Quantity 400 g
Formula Weight 74.922 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.