missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ART1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-91995-0.1ml
This item is not returnable.
View return policy
Description
ART1 Polyclonal antibody specifically detects ART1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ART1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| ADP-ribosyltransferase 1, ADP-ribosyltransferase 2, CD296, GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1, MGC133217, mono(ADP-ribosyl)transferase 1, RT6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 23-295 of human ART1 (NP_004305.2). QSHPITRRDLFSQEIQLDMALASFDDQYAGCAAAMTAALPDLNHTEFQANQVYADSWTLASSQWQERQARWPEWSLSPTRPSPPPLGFRDEHGVALLAYTANSPLHKEFNAAVREAGRSRAHYLHHFSFKTLHFLLTEALQLLGSGQRPPRCHQVFRGVHGLRFRPAGPRATVRLGGFASASLKHVAAQQFGEDTFFGIWTCLGAPIKGYSFFPGEEEVLIPPFETFQVINASRLAQGPARIYLRALGKHSTYNCEYIKDKKCKSGPCHLDNS | |
| 0.1 mL | |
| CD Markers, Immunology | |
| 417 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction