missing translation for 'onlineSavingsMsg'
Learn More

Artemether, MedChemExpress

Product Code. 16452376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452376 500 mg 500mg
16419076 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452376 Supplier Medchem Express Supplier No. HYN0402
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Artemether is an antimalarial for the treatment of resistant strains of falciparum malaria.

Artemether is an antimalarial for the treatment of resistant strains of falciparum malaria.

Specifications

Chemical Name or Material Artemether
Physical Form Solid
CAS 71963-77-4
Color Off-White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C16H26O5
For Use With (Application) COVID-19-immunoregulation
Synonym Dihydroqinghaosu methyl ether Dihydroartemisinin methyl ether SM224
Solubility Information DMSO : 100 mg/mL (335.15 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@H]1[C@@H](OC)O[C@@]2([H])[C@]34[C@@]([C@H](C)CC[C@]41[H])([H])CC[C@@](O2)(C)OO3
Molecular Weight (g/mol) 298.37
Formula Weight 298.37
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.