missing translation for 'onlineSavingsMsg'
Learn More

Artemisinin, MedChemExpress

Product Code. 16462006
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462006 500 mg 500mg
16401376 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462006 Supplier Medchem Express Supplier No. HYB0094
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Artemisinin (Qinghaosu), a sesquiterpene lactone, is an anti-malarial drug isolated from the aerial parts of Artemisia annua L. plants. Artemisinin inhibits AKT signaling pathway by decreasing pAKT in a dose-dependent manner. Artemisinin reduces cancer cell proliferation, migration, invasion, tumorigenesis and metastasis and has neuroprotective effects.

Artemisinin (Qinghaosu), a sesquiterpene lactone, is an anti-malarial drug isolated from the aerial parts of Artemisia annua L. plants. Artemisinin inhibits AKT signaling pathway by decreasing pAKT in a dose-dependent manner. Artemisinin reduces cancer cell proliferation, migration, invasion, tumorigenesis and metastasis and has neuroprotective effects.

Specifications

Chemical Name or Material Artemisinin
Physical Form Solid
CAS 63968-64-9
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C15H22O5
For Use With (Application) Cancer-Kinase/protease
Synonym Qinghaosu NSC 369397
Solubility Information DMSO : 50 mg/mL (177.10 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.35 mM; Need ultrasonic)
SMILES O=C1[C@H](C)[C@]2([H])CC[C@@H](C)[C@]3([H])CC[C@@](O4)(C)OO[C@]32[C@]4([H])O1
Molecular Weight (g/mol) 282.33
Formula Weight 282.33
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.