missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AS3MT Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92300-0.1ml
This item is not returnable.
View return policy
Description
AS3MT Polyclonal antibody specifically detects AS3MT in Mouse samples. It is validated for Western Blot
Specifications
| AS3MT | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| arsenic (+3 oxidation state) methyltransferase, arsenite methyltransferase, CYT19methyltransferase cyt19, EC 2.1.1.137, Methylarsonite methyltransferase, S-adenosyl-L-methionine:arsenic(III) methyltransferase, S-adenosylmethionine:arsenic (III) methyltransferase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 216-375 of human AS3MT (NP_065733.2). LAVLAQKIGFCPPRLVTANLITIQNKELERVIGDCRFVSATFRLFKHSKTGPTKRCQVIYNGGITGHEKELMFDANFTFKEGEIVEVDEETAAILKNSRFAQDFLIRPIGEKLPTSGGCSALELKDIITDPFKLAEESDSMKSRCVPDAAGGCCGTKKSC | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 57412 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction