missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASF1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519527
This item is not returnable.
View return policy
Description
This antibody is predicted to react with chicken and cow based on sequence homology. Store antibody at 4°C for 1-2 weeks. For long-term storage, store at -20°C.
This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The protein is a key component of a histone donor complex that functions in nucleosome assembly. It interacts with histones H3 and H4, and functions together with a chromatin assembly factor during DNA replication and repair.
Specifications
| ASF1A | |
| Polyclonal | |
| Unconjugated | |
| Q9Y294 | |
| ASF1A | |
| Synthetic peptide conjugated to KLH derived from within residues 150 to the C-terminus of Human ASF1 alpha/beta. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.2 mg/mL | |
| ASF1A | |
| RP3-329L24.1, CGI-98, CIA, DKFZp547E2110, HSPC146, CCG1-interacting factor A; anti-silencing function 1A; anti-silencing function protein 1 homolog A; hAsf1; hAsf1a; hCIA; histone chaperone ASF1A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25842 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction