missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASGPR1 Antibody (011), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89635PCP
This item is not returnable.
View return policy
Description
ASGPR1 Monoclonal antibody specifically detects ASGPR1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ASGPR1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 011 | |
| Immunohistochemistry-Paraffin | |
| ASGPR, ASGPR 1, ASGP-R 1, asialoglycoprotein receptor 1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ASGPR1 (Uniprot#: P07306; Gln62-Leu291). | |
| 0.1 mL | |
| Immunology, Virology Bacteria and Parasites | |
| 432 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction