Learn More
Invitrogen™ ASGR1 Recombinant Rabbit Monoclonal Antibody (103)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560404
 View more versions of this product
Product Code. 30536295
  • 429.00 € / 100µL
Description
ASGR1 Recombinant Monoclonal Antibody for ELISA
The asialoglycooprotein receptor (ASGPR) is a transmembrane glycoprotein (42 kDa) which mediates binding, internalization and degradation of extracellular glycoproteins that have exposed terminal galactose residues. The receptor is expressed on the surface of the hepatocytes in a polar manner, i. e. it is present on the sinusoidal and lateral plasma membranes but not on the bile canalicular membrane. The mammalian hepatic ASGPR mediate the endocytosis and degradation of serum proteins from which terminal sialic residues have been removed. The rat asialoglycoprotein receptor (ASGPR) consists of three polypeptide subunits: rat hepatic lectin (RHL)-1, RHL-2 and RHL-3. The monoclonal antibody 8D7 recognizes a subunit-specific epitope of RHL-1 of rat ASGPR. The monoclonal antibody 8D7 is cross reactive with human ASGPR.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ASGR1
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P34927
Asgr1
Recombinant Mouse ASGPR1/ASGR1 protein (Ser60-Asn284).
100 μL
Primary
Mouse
Antibody
IgG
ELISA
103
Unconjugated
Asgr1
ASGPR; ASGPR 1; ASGP-R 1; ASGPR1; ASGR; ASGR1; Asgr-1; asialoglycoprotein receptor (RHL1); asialoglycoprotein receptor 1; Asialoglycoprotein receptor 1 (hepatic lectin); asialoglycoprotein receptor RHL1; CLEC4H1; C-type lectin domain family 4 member H1; Hepatic lectin 1; hepatic lectin 1 RHL1; hepatic lectin 1, RHL1; hepatic lectin H1; HL-1; RATRHL1; RHL1; RHL-1; Unknown (protein for MGC:133691)
Rabbit
Protein A
RUO
11889
-20°C, Avoid Freeze/Thaw Cycles