missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asialoglycoprotein Receptor 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85578-25ul
This item is not returnable.
View return policy
Description
Asialoglycoprotein Receptor 2 Polyclonal specifically detects Asialoglycoprotein Receptor 2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Asialoglycoprotein Receptor 2 | |
| Polyclonal | |
| Western Blot 0.04 - 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:200-1:500 | |
| ASGPR 2, ASGP-R 2, ASGPR2, ASGP-R2, asialoglycoprotein receptor 2, CLEC4H2FLJ60040, C-type lectin domain family 4 member H2, HBxAg-binding protein, HBXBP, Hepatic lectin H2, HL-2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ASGR2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QSAQLQAELRSLKEAFSNFSSSTLTEVQAISTHGGSVGDKITSLGAKLEKQQQDLKADHDALLFHLKHFPVDLRFVACQMELLHSNGSQRTCC | |
| 25 μL | |
| Signal Transduction | |
| 433 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction