missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15491
This item is not returnable.
View return policy
Description
ASPA Polyclonal specifically detects ASPA in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ASPA | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| ACY-2, ACY2aminoacylase 2, Aminoacylase-2, ASPaminoacylase-2, aspartoacylase, aspartoacylase (aminoacylase 2, Canavan disease), EC 3.5.1.15 | |
| Rabbit | |
| 36 kDa | |
| 0.1 mL | |
| Neuroscience | |
| 443 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 1mg/mL | |
| Western Blot 1:5000-1:20000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Frozen | |
| P45381 | |
| ASPA | |
| Full length human Aspartoacylase Recombinant protein. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction