missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asparagine synthetase Antibody (S05-9E5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19507-50ul
This item is not returnable.
View return policy
Description
Asparagine synthetase Monoclonal antibody specifically detects Asparagine synthetase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Asparagine synthetase | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin | |
| asparagine synthetase, asparagine synthetase (glutamine-hydrolyzing), asparagine synthetase [glutamine-hydrolyzing], Cell cycle control protein TS11, EC 6.3.5.4, Glutamine-dependent asparagine synthetase, TS11, TS11 cell cycle control protein | |
| A synthetic peptide of human Asparagine synthetase (Uniprot # P08243) | |
| 50 μg | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 440 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S05-9E5 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction