Learn More
ASTM (ASTM D6045, D1500) Colour Reference Standard, Paragon Scientific™
ASTM Colour Reference Standards are for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.
Brand: Paragon Scientific 134010
Additional Details : Weight : 0.90000kg
Type | ASTM Color 3 |
---|
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to International Standards
- 500mL Pack Size supplied with MSDS
- 18 months shelf life
- Tamper evident security packaging
- Manufactured in the United kingdom
Chemical Identifiers
500 mL |
Specifications
500 mL | |
ASTM Color 3 |
Nominal values quoted