Learn More
ASTM (ASTM D6045, D1500) Colour Reference Standard, Paragon Scientific™
ASTM Colour Reference Standards are for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.
181.00 € - 194.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
16040152
|
Paragon Scientific
134000 |
ASTM Color 1 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050152
|
Paragon Scientific
134010 |
ASTM Color 3 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060152
|
Paragon Scientific
134020 |
ASTM Color 5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070152
|
Paragon Scientific
134030 |
ASTM Color 7 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050162
|
Paragon Scientific
134290 |
ASTM Color < 0.5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to International Standards
- 500mL Pack Size supplied with MSDS
- 18 months shelf life
- Tamper evident security packaging
- Manufactured in the United kingdom
Nominal values quoted