missing translation for 'onlineSavingsMsg'
Learn More

ASTM (ASTM D6045, D1500) Colour Reference Standard, Paragon Scientific™

Product Code. 16040152
Change view
Click to view available options
Unit Size:
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
16040152 500mL
16050152 500mL
16060152 500mL
16070152 500mL
16050162 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16040152 Supplier Paragon Scientific Supplier No. 134000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ASTM Colour Reference Standards are for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.

  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to International Standards
  • 500mL Pack Size supplied with MSDS
  • 18 months shelf life
  • Tamper evident security packaging
  • Manufactured in the United kingdom

Specifications

Type Color Reference Standard
Quantity 500 mL
compliance-icons
Product Identifier
  • ASTM Colour Reference Standard
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : white mineral oil

Nominal values quoted

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.