missing translation for 'onlineSavingsMsg'
Learn More

ASTM D4294, Curve #1, VHG™

Product Code. 17900446
Click to view available options
Quantity:
6 x 100 mL
Unit Size:
600mL
This item is not returnable. View return policy

Product Code. 17900446

Brand: VHG Labs ALKKTSMINW6CRV1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ASTM D4294, Curve #1
TRUSTED_SUSTAINABILITY

Specifications

Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 75 cSt Mineral Oil
Quantity 6 x 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.