missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM™ D6584 Standard Solution 3, Honeywell Fluka™
Shop All Honeywell Fluka Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
missing translation for 'unitSize'
1mL
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Specifications
| Packaging | Glass Bottle |
| CAS | 110-86-1, 111-03-5 |
| Density | 0.98 g/cm3 |
| Flash Point | 20°C |
| Physical Form | Liquid |
| Boiling Point | 115°C |
| Industry Type | Biofuels |
| Quantity | 1 mL |
| Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction