missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D4815/D5599 1,2-Dimethoxyethane IS, Dr. Ehrenstorfer

Product Code. 30313183
Change view
Click to view available options
Quantity:
5 x 2 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313183 5 x 2 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313183 Supplier Dr. Ehrenstorfer Supplier No. DREGS09010060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dimethoxyethane
CAS 110-71-4
Analyte or Component Names 1,2-Dimethoxyethane
Shipping Condition Room Temperature
Molecular Formula C4 H10 O2
Concentration 1000000 μg/mL
Certifications/Compliance ISO 17034
Synonym Ethane, 1,2-dimethoxy-, 1,2-Dimethoxyethane, 1,2-Ethanediol, dimethyl ether, 2,5-Dioxahexane, DME, DME (glycol ether), Dimethyl Cellosolve, Ethylene dimethyl ether, Ethylene glycol dimethyl ether, Glycol dimethyl ether, Glyme, Hisolve MMM, Methyl Monoglyme, Monoethylene glycol dimethyl ether, Monoglyme, NSC 60542, α,β-Dimethoxyethane
InChI Formula InChI=1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3
SMILES COCCOC
IUPAC Name 1,2-dimethoxyethane
Molecular Weight (g/mol) 90.12
Quantity 5 x 2 mL
Formula Weight 90.0681 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.