missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D4815/D5599 Internal Standard Mixture 5-10% in Oxygenate Free Gasoline, Dr. Ehrenstorfer

Product Code. 30313011
Change view
Click to view available options
Quantity:
60 x 2 mL
Unit Size:
120mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313011 60 x 2 mL 120mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313011 Supplier Dr. Ehrenstorfer Supplier No. DRESP09000237OG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dimethoxyethane∣Ethanol∣Regular Unleaded Gasoline
CAS 110-71-4, 64-17-5, 8006-61-9
Analyte or Component Names 1,2-Dimethoxyethane∣Ethanol∣Regular Unleaded Gasoline
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Oxygenate Free Gasoline
Industry Type Food Testing
Quantity 60 x 2 mL
Concentration or Composition (by Analyte or Components) 1,2-Dimethoxyethane 5 %∣Ethanol 10 %∣Regular Unleaded Gasoline 85 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.