missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D4815 Quantitative Peak ID Mixture 4.00-7.30 % (w/w), Dr. Ehrenstorfer

Product Code. 30316813
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316813 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316813 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000975

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dimethoxyethane∣1-Propanol∣2-Butanol∣Benzene∣Butyl Alcohol, n-butanol∣Diisopropyl ether∣Ethanol∣Ethyl tert-Butyl Ether (ETBE)∣Isobutanol∣Isopropyl alcohol∣Methanol∣Methyl tert-butyl ether∣Methylcyclopentane∣tert.-Butanol∣tert-Amyl Alcohol∣tert-Amyl Methyl Ether (TAME)
CAS 110-71-4, 71-23-8, 78-92-2, 71-43-2, 71-36-3, 108-20-3
Analyte or Component Names 1,2-Dimethoxyethane∣1-Propanol∣2-Butanol∣Benzene∣Butyl Alcohol, n-butanol∣Diisopropyl ether∣Ethanol∣Ethyl tert-Butyl Ether (ETBE)∣Isobutanol∣Isopropyl alcohol∣Methanol∣Methyl tert-butyl ether∣Methylcyclopentane∣tert.-Butanol∣tert-Amyl Alcohol∣tert-Amyl Methyl Ether (TAME)
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Industry Type Petrochemical Testing
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Diisopropyl ether 4 % (w/w)∣Ethyl tert-Butyl Ether (ETBE) 4 % (w/w)∣Methyl tert-butyl ether 4 % (w/w)∣Methylcyclopentane 4 % (w/w)∣Benzene 5 % (w/w)∣1,2-Dimethoxyethane 6 % (w/w)∣1-Propanol 7.3 % (w/w)∣2-Butanol 7.3 % (w/w)∣Butyl Alcohol, n-butanol 7.3 % (w/w)∣Ethanol 7.3 % (w/w)∣Isobutanol 7.3 % (w/w)∣Isopropyl alcohol 7.3 % (w/w)∣Methanol 7.3 % (w/w)∣tert.-Butanol 7.3 % (w/w)∣tert-Amyl Alcohol 7.3 % (w/w)∣tert-Amyl Methyl Ether (TAME) 7.3 % (w/w)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.