missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM Method D4815 Valve Timing Mixture, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Diisopropyl ether∣Ethyl tert-Butyl Ether (ETBE)∣Methyl tert-butyl ether∣Methylcyclopentane∣n-Hexane |
| CAS | 108-20-3, 637-92-3, 1634-04-4, 96-37-7, 110-54-3 |
| Analyte or Component Names | Diisopropyl ether∣Ethyl tert-Butyl Ether (ETBE)∣Methyl tert-butyl ether∣Methylcyclopentane∣n-Hexane |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Diisopropyl ether 10 wt%∣Ethyl tert-Butyl Ether (ETBE) 10 wt%∣Methyl tert-butyl ether 10 wt%∣n-Hexane 59.8 wt%∣Methylcyclopentane 100000 mg/kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?