missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D5580 Daily Quality Control Standard, Dr. Ehrenstorfer

Product Code. 30310306
Change view
Click to view available options
Quantity:
5 x 10 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310306 5 x 10 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310306 Supplier Dr. Ehrenstorfer Supplier No. DREGS0900076
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4,5-Tetramethylbenzene∣1,2,4-Trimethylbenzene∣2,2,4-Trimethylpentane∣Benzene∣Ethylbenzene∣Naphthalene∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Octane∣o-Xylene (1,2-Dimethylbenzene)∣p-Xylene (1,4-Dimethylbenzene)∣Toluene
CAS 95-93-2, 95-63-6, 540-84-1, 71-43-2, 100-41-4, 91-20-3
Analyte or Component Names 1,2,4,5-Tetramethylbenzene∣1,2,4-Trimethylbenzene∣2,2,4-Trimethylpentane∣Benzene∣Ethylbenzene∣Naphthalene∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Octane∣o-Xylene (1,2-Dimethylbenzene)∣p-Xylene (1,4-Dimethylbenzene)∣Toluene
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Quantity 5 x 10 mL
Concentration or Composition (by Analyte or Components) 1,2,4,5-Tetramethylbenzene 1 %∣Naphthalene 1 %∣n-Dodecane 1 %∣Ethylbenzene 2 %∣1,2,4-Trimethylbenzene 3 %∣Benzene 4 μg/mL∣n-Decane 10 %∣n-Octane 15 %∣2,2,4-Trimethylpentane 20 %∣n-Hexane 580 μg/mL∣Toluene 1780 μg/mL∣o-Xylene (1,2-Dimethylbenzene) 4340 μg/mL∣p-Xylene (1,4-Dimethylbenzene) 4340 μg/mL∣n-Heptane 10000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.