missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D5769 Quality Control Reference Material with 4 IS in Isooctane, Dr. Ehrenstorfer

Product Code. 30315412
Change view
Click to view available options
Quantity:
10 x 2 mL
Unit Size:
20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315412 10 x 2 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315412 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000134IO

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4,5-Tetramethylbenzene∣1,2,4-Trimethylbenzene∣2,2,4-Trimethylpentane∣Benzene∣Benzene-D6∣Ethylbenzene∣Ethylbenzene-d10∣m-Xylene (1,3-Dimethylbenzene)∣Naphthalene∣Naphthalene D8∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Octane∣o-Xylene (1,2-Dimethylbenzene)∣Toluene∣Toluene D8
CAS 95-93-2, 95-63-6, 540-84-1, 71-43-2, 1076-43-3, 100-41-4
Analyte or Component Names 1,2,4,5-Tetramethylbenzene∣1,2,4-Trimethylbenzene∣2,2,4-Trimethylpentane∣Benzene∣Benzene-D6∣Ethylbenzene∣Ethylbenzene-d10∣m-Xylene (1,3-Dimethylbenzene)∣Naphthalene∣Naphthalene D8∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Octane∣o-Xylene (1,2-Dimethylbenzene)∣Toluene∣Toluene D8
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Isooctane
Quantity 10 x 2 mL
Concentration or Composition (by Analyte or Components) Naphthalene D8 1 wt%∣m-Xylene (1,3-Dimethylbenzene) 3 wt%∣o-Xylene (1,2-Dimethylbenzene) 3 wt%∣n-Dodecane 5 wt%∣2,2,4-Trimethylpentane 12 wt%∣n-Decane 12 wt%∣n-Hexane 12 wt%∣n-Heptane 16.9 wt%∣n-Octane 16.9 wt%∣Benzene 10000 mg/kg∣Naphthalene 10000 mg/kg∣1,2,4,5-Tetramethylbenzene 20000 mg/kg∣Benzene-D6 20000 mg/kg∣Ethylbenzene-d10 20000 mg/kg∣1,2,4-Trimethylbenzene 30000 mg/kg∣Ethylbenzene 30000 mg/kg∣Toluene 90000 mg/kg∣Toluene D8 90000 mg/kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.