missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D6293 O-PONA Olefin Mixture, Dr. Ehrenstorfer

Product Code. 30318597
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318597 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318597 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000607HH
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Heptene∣1-Hexene∣1-Nonene∣1-Octene∣1-Pentene
CAS 592-76-7, 592-41-6, 124-11-8, 111-66-0, 109-67-1
Analyte or Component Names 1-Heptene∣1-Hexene∣1-Nonene∣1-Octene∣1-Pentene
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Hexane/Heptane
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) 1-Heptene 2 %∣1-Hexene 2 %∣1-Octene 2 %∣1-Nonene 3 %∣1-Pentene 5 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.