missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D7845 Check Standard 0.01-0.05 Wt% in Toluene, Dr. Ehrenstorfer

Product Code. 30319772
Change view
Click to view available options
Quantity:
10 x 2 mL
Unit Size:
20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319772 10 x 2 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319772 Supplier Dr. Ehrenstorfer Supplier No. DREGS09001085TO

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (+)-Limonene∣(1R)-(+)-beta-Pinene∣1-Methylnaphthalene∣1-Phenoxy-2-propanol∣1-Phenylethanol∣2,4-Dimethylphenol∣2,4-dimethylstyrene∣2,5-dimethylstyrene∣2-Ethylphenol∣2-Methylnaphthalene∣2-Methylstyrene∣2-Phenoxy-1-propanol∣2-Phenylethanol∣3-Ethylphenol∣3-Methylstyrene∣4-Ethylphenol∣4-Isopropylphenol∣4-Methylstyrene∣Acrylic acid butyl ester∣alpha-Methylstyrene∣Butyl Alcohol, n-butanol∣Cyclohexanol∣Dicyclopentadiene∣Di-n-butylether∣DL-alpha-Pinene∣Ethylbenzene-d10∣Indene∣Naphthalene∣p,a-Dimethylstyrene∣Phenol∣Phenoxyethanol∣Styrene∣Styrene Glycol∣trans-beta-Methylstyrene
CAS 5989-27-5, 19902-08-0, 90-12-0, 770-35-4, 98-85-1, 105-67-9
Analyte or Component Names (+)-Limonene∣(1R)-(+)-beta-Pinene∣1-Methylnaphthalene∣1-Phenoxy-2-propanol∣1-Phenylethanol∣2,4-Dimethylphenol∣2,4-dimethylstyrene∣2,5-dimethylstyrene∣2-Ethylphenol∣2-Methylnaphthalene∣2-Methylstyrene∣2-Phenoxy-1-propanol∣2-Phenylethanol∣3-Ethylphenol∣3-Methylstyrene∣4-Ethylphenol∣4-Isopropylphenol∣4-Methylstyrene∣Acrylic acid butyl ester∣alpha-Methylstyrene∣Butyl Alcohol, n-butanol∣Cyclohexanol∣Dicyclopentadiene∣Di-n-butylether∣DL-alpha-Pinene∣Ethylbenzene-d10∣Indene∣Naphthalene∣p,a-Dimethylstyrene∣Phenol∣Phenoxyethanol∣Styrene∣Styrene Glycol∣trans-beta-Methylstyrene
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Toluene
Quantity 10 x 2 mL
Concentration or Composition (by Analyte or Components) (+)-Limonene 0.1 wt %∣(1R)-(+)-beta-Pinene 0.1 wt %∣1-Methylnaphthalene 0.1 wt %∣1-Phenoxy-2-propanol 0.1 wt %∣1-Phenylethanol 0.1 wt %∣2,4-Dimethylphenol 0.1 wt %∣2,4-dimethylstyrene 0.1 wt %∣2,5-dimethylstyrene 0.1 wt %∣2-Ethylphenol 0.1 wt %∣2-Methylnaphthalene 0.1 wt %∣2-Methylstyrene 0.1 wt %∣2-Phenoxy-1-propanol 0.1 wt %∣2-Phenylethanol 0.1 wt %∣3-Ethylphenol 0.1 wt %∣3-Methylstyrene 0.1 wt %∣4-Ethylphenol 0.1 wt %∣4-Isopropylphenol 0.1 wt %∣4-Methylstyrene 0.1 wt %∣Acrylic acid butyl ester 0.1 wt %∣alpha-Methylstyrene 0.1 wt %∣Butyl Alcohol, n-butanol 0.1 wt %∣Cyclohexanol 0.1 wt %∣Dicyclopentadiene 0.1 wt %∣Di-n-butylether 0.1 wt %∣DL-alpha-Pinene 0.1 wt %∣Ethylbenzene-d10 0.1 wt %∣Indene 0.1 wt %∣Naphthalene 0.1 wt %∣p,a-Dimethylstyrene 0.1 wt %∣Phenol 0.1 wt %∣Phenoxyethanol 0.1 wt %∣Styrene 0.1 wt %∣Styrene Glycol 0.1 wt %∣trans-beta-Methylstyrene 0.1 wt %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.