missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Astrocytomas Antibody (J1-31), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29820-0.025ML
This item is not returnable.
View return policy
Description
Astrocytomas Monoclonal specifically detects Astrocytomas in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Astrocytomas | |
| Monoclonal | |
| Unconjugated | |
| Ascites with No Preservative | |
| Mouse | |
| Unpurified | |
| RUO | |
| The antibody recognizes an intracellular protein antigen (MW 30 kDa) expressed by human and rat astrocytes and other specialized glia (Muller cells of the retina, Bergmann fibers of the cerebellar cortex, tanycytes of the hypothalamus and ciliated ependymal cells) in the central nervous system (CNS).The antibody has recently been found to be a specific marker for low grade astrocytoma in human brain tissue. The antibody is able to distinguish between low grade astrocytoma and normal reactive gliosis (patent application filed). Monoclonal antibody J1-31 was raised against crude homogenate of brain tissue from a multiple sclerosis (MS) patient (autopsy sample; Malhotra et al.: Microbios Letters 26:151-157, 1984). In human brain, MAb J1-31 recognizes an intracellular protein antigen (J1-31 antigen), which bands at approximately 30,000 daltons under reducing conditions for sodium dodecyl sulfate gel electrophoresis (Singh et al.: Bioscience Reports 6:73-79, 1986). By immunofluorescence microscopy, MAb J1-31 stains those cells that are also stained by antiserum to glial fibrillary acidic protein (GFAP), namely astrocytes, retinal Muller cells, and tanycytes in the ependyma (Predy et al.: Bioscience Reports 7:491-502, 1987). In addition, MAb J1-31 stains ciliated ependymal cells that do not express GFAP (Malhotra, SK (1989) J Neurosci Res. 22(1):36-49). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgM |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
| J1-31 | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Astrocytoma Marker, Astrocytomas Marker, Low Grade Astrocytoma Marker | |
| Human cerebral white matter plaque materials from a multiple sclerosis patient. | |
| 0.025 mL | |
| Primary | |
| Human, Rat | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction