missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asymmetric dimethylarginine, MedChemExpress
Description
Asymmetric dimethylarginine is an endogenous inhibitor of nitric oxide synthase (NOS), and functions as a marker of endothelial dysfunction in a number of pathological states.
Specifications
Specifications
| Chemical Name or Material | Asymmetric dimethylarginine |
| Physical Form | Powder |
| CAS | 30315-93-6 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C8H18N4O2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | H2O : 100 mg/mL (494.44 mM; adjust pH to 2-3 with HCl) ∣DMSO : 100 mg/mL (494.44 mM; Need ultrasonic) |
| SMILES | CN(C)C(NCCC[C@H](N)C(O)=O)=N |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?