missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATAD2 Antibody (CL0182), Novus Biologicals™
Mouse Monoclonal Antibody
369.00 € - 500.00 €
Specifications
| Antigen | ATAD2 |
|---|---|
| Clone | CL0182 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18405221
|
Novus Biologicals
NBP2-14786-25ul |
25ul |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18147301
|
Novus Biologicals
NBP2-14786 |
0.1 mL |
500.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATAD2 Monoclonal specifically detects ATAD2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| ATAD2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Protein Phosphatase | |
| AAA nuclear coregulator cancer-associated protein, ANCCAMGC131938, ATPase family AAA domain-containing protein 2, ATPase family, AAA domain containing 2, DKFZp667N1320, EC 3.6.1.3, MGC142216, MGC29843, MGC5254, PRO2000 | |
| ATAD2 | |
| IgG1 | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL0182 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 29028 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: TAYAIIKEELDEDFEQLCEEIQESRKKRGCSSSKYAPSYYHVMPKQNSTLVGDKRSDPEQNEKLKTPSTPVACSTPAQLKRKIRKKSNWYLGTIKKRRKISQAKDDSQNAIDHK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title