missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATAD2 Monoclonal Antibody (952947)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA562001

Product Code. 30534454

  • 429.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

ATAD2 is a member of the ATPase family of proteins. Members of this family share a conserved region of around 220 amino acids that contain an ATP-binding site. They also contain one or two AAA (ATPases Associated with Diverse Cellular Activities) domains. ATPase family proteins perform chaperone-like functions that assist in assembly, operation, and disassembly of protein complexes. Involved in the estrogen-induced cell proliferation and cell cycle progression of breast cancer cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATAD2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q6PL18
ATAD2
Human ATAD2 synthetic peptide.
100 μg
Primary
Human
Antibody
IgG2b
Immunocytochemistry
952947
Unconjugated
ATAD2
2610509G12Rik; AAA nuclear coregulator cancer-associated protein; ANCCA; Atad2; ATPase family AAA domain-containing protein 2; ATPase family, AAA domain containing 2; CT137; L16; PRO2000
Mouse
Protein A/G
RUO
29028
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.