missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ataxin-2-like protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48741
This item is not returnable.
View return policy
Description
Ataxin-2-like protein Polyclonal antibody specifically detects Ataxin-2-like protein in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Specifications
| Ataxin-2-like protein | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated | |
| A2DA2RP, A2LG, A2lp, ataxin 2 related protein, ataxin 2-like, Ataxin-2 domain protein, ataxin-2-like protein, Ataxin-2-related protein | |
| This antibody was developed against a recombinant protein corresponding to amino acids: APISASCPEPPIGSAVPTSSASIPVTSSVSDPGVGSISPASPKISLAPTDVKELSTKEPGRTLEPQELARIAGKVPGLQNEQKRFQLEELRK | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 11273 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction