missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATBF1/ZFHX3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 624.00 €
Specifications
| Antigen | ATBF1/ZFHX3 |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18447811
|
Novus Biologicals
NBP2-34126-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18160215
|
Novus Biologicals
NBP2-34126 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATBF1/ZFHX3 Polyclonal specifically detects ATBF1/ZFHX3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ATBF1/ZFHX3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| Q15911 | |
| 463 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KAQVQVPQQSHQQILPQQQQNQLSIAQSHSALLQPSQHPEKKNKLVIKEKEKESQRERDSAEGGEGNTGPKETLPDA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Prostate Cancer | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Alpha-fetoprotein enhancer-binding protein, AT motif-binding factor, AT motif-binding factor 1, ATBF1alpha-fetoprotein enhancer binding protein, AT-binding transcription factor 1ATBT, ZFH-3, zinc finger homeobox 3, zinc finger homeobox protein 3, Zinc finger homeodomain protein 3, ZNF927 | |
| ZFHX3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title