missing translation for 'onlineSavingsMsg'
Learn More

Atenolol, 98%

Product Code. 13565040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13565040 5 g Glass bottle 5g
13555040 1 g Glass bottle 1g
13575040 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13565040 Supplier Thermo Scientific Acros Supplier No. 449550050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29122-68-7 | C14H22N2O3 | 266.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29122-68-7
Molecular Formula C14H22N2O3
Molecular Weight (g/mol) 266.34
InChI Key METKIMKYRPQLGS-UHFFFAOYSA-N
Synonym atenolol, tenormin, tenormine, normiten, blokium, prenormine, tenoblock, atehexal, betablok, cuxanorm
PubChem CID 2249
ChEBI CHEBI:2904
IUPAC Name 2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide
SMILES CC(C)NCC(COC1=CC=C(C=C1)CC(=O)N)O

Specifications

Melting Point 146.0°C to 148.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Merck Index 15,85
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol, acetic acid, dmso, ethanol, insoluble in acetonitrile, ethylacetate, chlorofor, m
Formula Weight 266.34
Percent Purity 98%
Chemical Name or Material Atenolol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.