missing translation for 'onlineSavingsMsg'
Learn More

Atenolol, MedChemExpress

Product Code. p-200096272
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439847 5 g 5g
16411689 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16439847 Supplier Medchem Express Supplier No. HY17498

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Atenolol ((RS)-Atenolol) is a cardioselective β1-adrenergic receptor blocker, with a Ki of 697 nM atβ1-adrenoceptor in guine pig left ventricle membrane. Atenolol can be used for the research of hypertension and angina pectoris.

Atenolol ((RS)-Atenolol) is a cardioselective β1-adrenergic receptor blocker, with a Ki of 697 nM atβ1-adrenoceptor in guine pig left ventricle membrane. Atenolol can be used for the research of hypertension and angina pectoris.

Specifications

Chemical Name or Material Atenolol
CAS 29122-68-7
Color White
Purity Grade Notes Research
Molecular Formula C14H22N2O3
Quantity 5 g
Synonym (RS)-Atenolol
Solubility Information DMSO : 100 mg/mL (375.46 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (31.28 mM; Need ultrasonic)
SMILES O=C(N)CC1=CC=C(OCC(O)CNC(C)C)C=C1
Molecular Weight (g/mol) 266.34
Formula Weight 266.34
Percent Purity 99.61%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.