Learn More
ATF7 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29770-1-AP-150UL
Description
The ATF7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ATF7. This antibody recognizes human, mouse, rat antigen. The ATF7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| ATF7 | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| ATF7, ATFA, Transcription factor ATF A | |
| 150 μL | |
| 11016 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |