missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG10 Antibody (SR1389), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21730-50ul
This item is not returnable.
View return policy
Description
ATG10 Monoclonal antibody specifically detects ATG10 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| ATG10 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1389 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| APG10, APG10 autophagy 10-like (S. cerevisiae), APG10L, APG10-like, ATG10 autophagy related 10 homolog (S. cerevisiae), Autophagy-related protein 10, DKFZp586I0418, EC 6.3.2.-, FLJ13954, pp12616, ubiquitin-like-conjugating enzyme ATG10 | |
| A synthesized peptide derived from human ATG10 (Uniprot #: Q9H0Y0) | |
| 50 μg | |
| Autophagy | |
| 83734 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction