Learn More
Invitrogen™ ATG4A Recombinant Rabbit Monoclonal Antibody (23GB3235), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552603
 View more versions of this product
Product Code. 30089633
  • 186.00 € / 20µL
Description
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some malignant tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Transcript variants that encode distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATG4A
Recombinant Monoclonal
Unconjugated
ATG4A
AI627006; APG4 (ATG4) autophagy-related homolog A; APG4 autophagy 4 homolog A; APG4A; ATG4 autophagy related 4 homolog A; Atg4a; Atg4al; Atg4al1; AUTL2; AUT-like 2 cysteine endopeptidase; AUT-like 2, cysteine endopeptidase; autophagin 2; autophagin-2; autophagy 4 homolog A; autophagy related 4 homolog A; autophagy related 4A cysteine peptidase; autophagy related 4A, cysteine peptidase; autophagy related 4A, cysteine peptidase-like 1; autophagy-related 4A; autophagy-related cysteine endopeptidase 2; Autophagy-related protein 4 homolog A; AV169859; cysteine protease APG4A; cysteine protease ATG4A; hAPG4A; MGC107179
Rabbit
Affinity Chromatography
RUO
115201, 666468, 678769
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB3235
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8C9S8, Q8WYN0
ATG4A
A synthetic peptide of human Atg4A (1-84AA).
20 μL
Primary
Human, Mouse, Rat