missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
379.00 € - 563.00 €
Specifications
| Antigen | ATN1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18489571
|
Novus Biologicals
NBP1-90044-25ul |
25 μL |
379.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18035204
|
Novus Biologicals
NBP1-90044 |
0.1 mL |
563.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATN1 Polyclonal specifically detects ATN1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ATN1 | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1822 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:HPYAMSPSLGSLRPYPPGPAHLPPPHSQVSYSQAGPNGPPVSSSSNSSSSTSQGSYPCSHPSPSQGPQGAPYPFPPVPTVTTSSATLSTVIATVASSPAGYKTASPPGPPPYGKRAPSPGAYKTATPPGYKPGSPPSFRTGTPPGY | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| atrophin 1, atrophin-1, B37, D12S755E, dentatorubral-pallidoluysian atrophy (atrophin-1), Dentatorubral-pallidoluysian atrophy protein, DRPLANOD, HRS | |
| ATN1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title