Learn More
Invitrogen™ ATP1A1 Monoclonal Antibody (M7-PB-E9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA3928
Description
MA3-928 detects sodium/potassium ATPase alpha from human, mouse, bovine, sheep, canine, chicken and porcine tissues. This antibody detects the alpha 3 isoform in rat only. MA3-928 has been successfully used in Western blot, immunohistochemistry, immunoprecipitation, immunocytochemistry, immunofluorescence, and ELISA procedures. By Western blot, this antibody detects an ~110 kDa protein representing alpha sodium/potassium ATPase from MDCK cell extract. Immunohistochemical staining of alpha sodium/potassium ATPase in porcine heart with MA3-928 yields a pattern consistent with the plasma membrane localization. The MA3-928 antigen is purified sheep kidney alpha sodium/potassium ATPase. This antibody recognizes an epitope between amino acid residues 646 and 652 of the sheep kidney alpha sodium/potassium ATPase.
The protein encoded by this gene belongs to the family of P-type cation transport ATPases, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The catalytic subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes an alpha 1 subunit. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| ATP1A1 | |
| Monoclonal | |
| 1.87 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| P04074, P05023, P05024, P06685, P09572, P50997, Q08DA1, Q8VDN2 | |
| ATP1A1 | |
| Purified alpha sodium/potassium ATPase from sheep kidney. | |
| 400 μL | |
| Primary | |
| Bovine, Canine, Chicken, Human, Mouse, Sheep, Pig, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| M7-PB-E9 | |
| Unconjugated | |
| ATP1A1 | |
| (Na+ + K+)-ATPase; (Na+, K+)-ATPase alpha-subunit; alpha subunit of sodium potassium ATPase; atp1a1; atp1a1-a; atp1a1a.4; atp1a1-b; ATP1B; Atpa-1; ATPase; ATPase Na+/K+ transporting subunit alpha 1; ATPase, Na+/K+ transporting, alpha 1 polypeptide; ATPase, Na+K+ transporting, alpha 1; BC010319; K-ATPase; MGC1798; Na(+)/K(+) ATPase alpha-1 subunit; Na, K-ATPase alpha-1 subunit; Na, K-ATPase, alpha-A catalytic polypeptide; Na,K-ATPase alpha-1 subunit; Na,K-ATPase catalytic subunit alpha-A protein; Na/K ATPase alpha 1 isoform; Na/K ATPase alpha 1 subunit; Na+ ,K+ -ATPase alpha subunit; Na+ /K+ ATPase alpha 1 subunit; Na+, K+-ATPase beta-subunit precursor; Na+/K+ ATPase; Na+/K+ ATPase 1; Na+/K+ ATPase Alpha; Na+/K+ -ATPase alpha 1 subunit; Na+/K+ transporting alpha 1 polypeptide; Na+K+ ATPase; Na+K+ATPase; Nkaa1b; RP4-655J12.1; sarcolemmal sodium/potassium-ATPase alpha 1; Sodium Potassium ATPase; sodium pump 1; sodium pump subunit alpha-1; sodium/potassium-ATPase alpha-subunit 1; sodium/potassium-transporting ATPase alpha-1 chain; sodium/potassium-transporting ATPase subunit alpha-1; sodium-potassium ATPase catalytic subunit alpha-1; sodium-potassium-ATPase, alpha 1 polypeptide; unnamed protein product | |
| Mouse | |
| Protein A | |
| RUO | |
| 11928, 24211, 282144, 396530, 397481, 403992, 443381, 476 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.