missing translation for 'onlineSavingsMsg'
Learn More

ATP1A1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16846675 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
Unit Size:
150µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16846675 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16846675 Supplier Proteintech Supplier No. 551871AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is specific to ATP1A1.

Na,K-ATPase is an integral membrane protein complex that hydrolyzes ATP to maintain the transmembrane gradients of Na+ and K+ found in most mammalian cells. The enzyme is comprised of an alpha and beta subunit. The alpha-polypeptide has been shown to be the catalytically active subunit, wherease the Beta-polypeptide appears to be necessary for the assembly and transport of the sodium pump to the plasma membrane. Na,K-ATPase alpha-1 subunit, in brain, is expressed in both neuronal and non-neuronal cells.

Specifications

Antigen ATP1A1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATP1A1
Gene Accession No. P05023, Q8VDN2
Gene Alias ATP1A1, Na(+)/K(+) ATPase 1, Na(+)/K(+) ATPase alpha 1 subunit, Na+/K+ ATPase 1
Gene Symbols ATP1A1
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11928, 476
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.