missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V0A1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92219-0.1ml
This item is not returnable.
View return policy
Description
ATP6V0A1 Polyclonal antibody specifically detects ATP6V0A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| ATP6V0A1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| a1, ATP6N1, ATP6N1AATPase, H+ transporting, lysosomal non-catalytic accessory protein 1(110/116kD), ATP6V0, ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalyticaccessory protein 1A (110/116kD), ATPase, H+ transporting, lysosomal V0 subunit a isoform 1, ATPase, H+ transporting, lysosomal V0 subunit a1, Clathrin-coated vesicle/synaptic vesicle proton pump 116 kDa subunit, DKFZp781J1951, Stv1, Vacuolar adenosine triphosphatase subunit Ac116, Vacuolar proton pump subunit 1, vacuolar proton pump, subunit 1, vacuolar proton translocating ATPase 116 kDa subunit A, Vacuolar proton translocating ATPase 116 kDa subunit a isoform 1, vacuolar-type H(+)-ATPase 115 kDa subunit, V-ATPase 116 kDa, V-ATPase 116 kDa isoform a1, Vph1, VPP1H(+)-transporting two-sector ATPase, 116 kDa accessory protein A1, V-type proton ATPase 116 kDa subunit a, V-type proton ATPase 116 kDa subunit a isoform 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-300 of human ATP6V0A1 (NP_001123493.1). EMDRKLRFVEKEIRKANIPIMDTGENPEVPFPRDMIDLEANFEKIENELKEINTNQEALKRNFLELTELKFILRKTQQFFDEMADPDLLEESSSLLEPSEMGRGTPLRLGFVAGVINRERIPTFERMLWRVCRGNVFLRQAEIENPLEDPVTGDYVHKSVFIIFFQGDQLKNRVKKICEGFRASLYPCPETPQERKEMASGVNTRIDDLQMVLNQTEDHRQRVLQAAAKNIRVWFIKVRKM | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 535 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction