missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP6V1D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117802
This item is not returnable.
View return policy
Description
ATP6V1D Polyclonal Antibody for Western Blot, IHC (P), IP
This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c, and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene encodes the V1 domain D subunit protein.
Specifications
| ATP6V1D | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9Y5K8 | |
| Atp6v1d | |
| E. coli-derived Human ATP6V1D/Atp6m fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Atp6v1d | |
| 1110004P10Rik; ATP6M; ATP6V1D; ATPase H+ transporting V1 subunit D; ATPase, H+ transporting lysosomal (vacuolar proton pump); ATPase, H+ transporting lysosomal, member M; ATPase, H+ transporting, lysosomal (vacuolar proton pump); ATPase, H+ transporting, lysosomal 34kD, V1 subunit D; ATPase, H+ transporting, lysosomal 34kDa, V1 subunit D; ATPase, H+ transporting, lysosomal V1 subunit D; ATPase, H+ transporting, V1 subunit D; H(+)-transporting two-sector ATPase, subunit M; lysosomal 34kDa; vac; vacuolar ATP synthase subunit D; vacuolar H-ATPase subunit D; vacuolar proton pump D subunit; vacuolar proton pump delta polypeptide; vacuolar proton pump subunit D; vacuolar proton-ATPase subunit D; VATD; V-ATPase 28 kDa accessory protein; V-ATPase D subunit; V-ATPase subunit D; VMA8; V-type proton ATPase subunit D | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 51382 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction