missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPase Na+/K+ beta 3 Antibody (S02-1B4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19946-50ul
This item is not returnable.
View return policy
Description
ATPase Na+/K+ beta 3 Monoclonal antibody specifically detects ATPase Na+/K+ beta 3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ATPase Na+/K+ beta 3 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S02-1B4 | |
| Unconjugated | |
| ATPase, Na+/K+ transporting, beta 3 polypeptide, ATPB-3, CD298, CD298 antigen, FLJ29027, Na, K-ATPase beta-3 polypeptide, sodium/potassium-dependent ATPase beta-3 subunit, Sodium/potassium-dependent ATPase subunit beta-3, sodium/potassium-transporting ATPase beta-3 chain, sodium/potassium-transporting ATPase subunit beta-3 | |
| A synthetic peptide of human ATPase Na+/K+ beta 3 (Uniprot # P54709) | |
| 50 μg | |
| Cancer, CD Markers, Endocrinology, Immunology, Signal Transduction | |
| 483 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction